α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline
185690100 185690500
ACRO185690100EA 30.7 EUR
ACRO185690100 ACRO185690500
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98+%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Formule: C₈H₅F₆N
Kookpunt: 85 °C (15 mmHg)
Smeltpunt: 3 °C
Dichtheid: 1,47 g/cm³
Vlampunt: 83 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000394
CAS nummer: 328-74-5
EINECS: 206-335-0
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear light yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
Titration with HClO4 ≥98.0 %