3,5-Dimethylphenyl isocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01800.1 SB01800.10 SB01800.50
MAYBSB01800.1EA 0 EUR
MAYBSB01800.1 MAYBSB01800.10 MAYBSB01800.50
3,5-Dimethylphenyl isocyanate
3,5-Dimethylphenyl isocyanate
Formule: C₉H₉NO
Kookpunt: 205 °C (1013 hPa)
Dichtheid: 1,04 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00013868
CAS nummer: 54132-75-1
EINECS: 258-987-0
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR