2,5-Dimethylphenyl isocyanate ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L15999.03
L15999.06
L15999.03EA
24.4
EUR
L15999.03
L15999.06
2,5-Dimethylphenyl isocyanate ≥97%
2,5-Dimethylphenyl isocyanate
Purity: 97%
CAS No.: 40397-98-6
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 90°(194°F)
Refractive Index: 1.5315
Boiling Point: 210°
Beilstein Reference: 1100841
UN No.: 2206
MDL No.: MFCD00013854
Formule:
C₉H₉NO Kookpunt: 210 °C Dichtheid: 1,043 Vlampunt: 90 °C (194 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00013854 CAS nummer: 40397-98-6 UN: 2206 ADR: 6.1,II |