2,6-Xylyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2,6-Dimethylphenylisothiocyanate, 2,6-Dimethylphenyl isothiocyanate

SB01642.1 SB01642EA SB01642.50
MAYBSB01642.1EA 0 EUR
MAYBSB01642.1 MAYBSB01642EA MAYBSB01642.50
2,6-Xylyl isothiocyanate
2,6-Xylyl isothiocyanate
Formule: C₉H₉NS
MW: 163.24 g/mol
Kookpunt: 67 °C (0.4 mmHg)
Dichtheid: 1.085 g/cm³ (20 °C)
MDL Nummer: MFCD00038715
CAS nummer: 19241-16-8
EINECS: 242-906-0
UN: 3334

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR