6-Nitro-2,3-xylidine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2,3-Dimethyl-6-nitroaniline

SB00708.1 SB00708.10 SB00708.50
MAYBSB00708.1EA 0 EUR
MAYBSB00708.1 MAYBSB00708.10 MAYBSB00708.50
6-Nitro-2,3-xylidine
6-Nitro-2,3-xylidine
Formule: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Smeltpunt: 120
MDL Nummer: MFCD00010148
CAS nummer: 59146-96-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR