2,6-Xylenol, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar


M-Clarity™ quality level = MQ100

36715-1GEA 33.3 EUR
36715-1G
2,6-Xylenol, Supelco®
2,6-Xylenol
Formule: C₈H₁₀O
MW: 122,17 g/mol
Kookpunt: 203 °C (1013 hPa)
Smeltpunt: 41…42 °C
Dichtheid: 1,13 g/cm³ (20 °C)
Vlampunt: 73 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002240
CAS nummer: 576-26-1
EINECS: 209-400-1
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR