Thymol 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
150331000 150335000 150330025
ACRO150331000EA 50.5 EUR
ACRO150331000 ACRO150335000 ACRO150330025
Thymol 99%
Thymol
Formule: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
Kookpunt: 232 °C (760 mmHg)
Smeltpunt: 48…52 °C
Dichtheid: 0,96 g/cm³
Vlampunt: 102 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002309
CAS nummer: 89-83-8
EINECS: 201-944-8
UN: 2430
ADR: 8,III
Merck Index: 13,09476
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b3268b34e160525798cab810087412f9b9a9db6&vid=7293cafb537e97291a403e856ca7fdb0223ba714&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥98.5 %
Residue after evaporation ≤0.05 %