Thymol 99.01-101.0% BP, NF, Analytical reagent (AR), Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
16254-100G 16254-500G
16254-100GEA 49.3 EUR
16254-100G 16254-500G
Thymol 99.01-101.0% BP, NF, Analytical reagent (AR), Sigma-Aldrich®
Thymol
Thymol meets analytical specification of Ph. Eur., BP, NF, 99 to 101%.
Formule: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
Kookpunt: 233 °C (1013 hPa)
Smeltpunt: 49…51 °C
Dichtheid: 0,965 g/cm³ (25 °C)
Vlampunt: 104 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002309
CAS nummer: 89-83-8
EINECS: 201-944-8
UN: 2430
ADR: 8,III
Merck Index: 13,09476
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b3268b34e160525798cab810087412f9b9a9db6&vid=7293cafb537e97291a403e856ca7fdb0223ba714&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99 - 101%
Residue after evaporation ≤0.05%