4-Methylbenzophenone 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
126350500 126352500
ACRO126350500EA 57.4 EUR
ACRO126350500 ACRO126352500
4-Methylbenzophenone 97%
4-Methylbenzophenone
Formule: C₁₄H₁₂O
Kookpunt: 326 °C (1013 hPa)
Smeltpunt: 53…57 °C
Dichtheid: 0,9926 g/cm³ (20 °C)
Vlampunt: 143 °C
MDL Nummer: MFCD00008553
CAS nummer: 134-84-9
EINECS: 205-159-1
Merck Index: 12,07470
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥96.0 %