2-Iodomesitylene
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2,4,6-Trimethyliodobenzene, 1-Iodo-2,4,6-trimethylbenzene, 2-Iodo-1,3,5-trimethylbenzene, Iodomesitylene
CD07694.1 CD07694.50
MAYBCD07694.1EA 0 EUR
MAYBCD07694.1 MAYBCD07694.50
2-Iodomesitylene
2-Iodomesitylene
Formule: C₉H₁₁I
MW: 246.09 g/mol
MDL Nummer: MFCD00013707
CAS nummer: 4028-63-1
EINECS: 223-709-9