2,4,6-Triisopropylbenzenesulphonyl chloride 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2,4,6-Triisopropylbenzene-1-sulphonyl chloride
152790250 152791000
ACRO152790250EA 51.6 EUR
ACRO152790250 ACRO152791000
2,4,6-Triisopropylbenzenesulphonyl chloride 97%
2,4,6-Triisopropylbenzenesulphonyl chloride
Formule: C₁₅H₂₃ClO₂S
MW: 302.87 g/mol
Smeltpunt: 93…97 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007433
CAS nummer: 6553-96-4
EINECS: 229-479-6
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance White to light beige Crystalline solid
Infrared spectrum Conforms
Melting point 93 to 97 °C
Titration Argentometric ≥96.0 %
Gel strength ≥700 g/cm² (1.5 %)
Gel point 32 to 38 °C (1.5 %)
Ash ≤5 %
pH 6.0 to 7.5 (10 % suspension)