5-tert-Butyl-m-xylene ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-tert-Butyl-3,5-dimethylbenzene, 1-(tert-Butyl)-3,5-dimethylbenzene
A19220.22 A19220.36
A19220.22EA 29.6 EUR
A19220.22 A19220.36
5-tert-Butyl-m-xylene ≥98%
5-tert-Butyl-m-xylene
Purity: 98% CAS No.: 98-19-1 Molecular Formula: C12H18 Formula Weight: 162.27 Flash Point: 84°(183°F) Refractive Index: 1.4980 Melting Point: -22° Boiling Point: 207-209° Beilstein Reference: 1853314 MDL No.: MFCD00008832
Formule: C₁₂H₁₈
MW: 162.27 g/mol
Kookpunt: 207…209 °C
Smeltpunt: –22 °C
Dichtheid: 0.86
Vlampunt: 84 °C (183 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008832
CAS nummer: 98-19-1
EINECS: 202-647-6
Order Now
Specificaties
Environmentally Preferable