4-Biphenylcarbonyl chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
171250100 171250500
ACRO171250100EA 49.3 EUR
ACRO171250100 ACRO171250500
4-Biphenylcarbonyl chloride 98%
4-Biphenylcarbonyl chloride
Formule: C₁₃H₉ClO
Kookpunt: 195…200 °C (5 mmHg)
Smeltpunt: 110…114 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000692
CAS nummer: 14002-51-8
EINECS: 237-804-8
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 114 °C
Titration Argentometric ≥97.5 % (ex Cl)