Bifenyl-4-ol 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
158710010 158712500
ACRO158710010EA 180 EUR
ACRO158710010 ACRO158712500
Bifenyl-4-ol 97%
Bifenyl-4-ol
Formule: C₁₂H₁₀O
Kookpunt: 321 °C (1013 hPa)
Smeltpunt: >163 °C
Dichtheid: 1,29 g/cm³ (20 °C)
Vlampunt: 165 °C
MDL Nummer: MFCD00002347
CAS nummer: 92-69-3
EINECS: 202-179-2
UN: 3077
Merck Index: 12,07459
Order Now
Specification Test Results
Appearance White to beige Flakes
Infrared spectrum Conforms
Melting point 163 to 168 °C
GC ≥96.0 %
Residue after reduction ≤0.02 %