Coumarin ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone
C0395-25G C0395-500G
TCIAC0395-25GEA 19.8 EUR
TCIAC0395-25G TCIAC0395-500G
Coumarin ≥99.0% (by GC)
Coumarin
Formule: C₉H₆O₂
Kookpunt: 298 °C (1013 hPa)
Smeltpunt: 67…69 °C
Dichtheid: 0,935 g/cm³ (20 °C)
Vlampunt: 162 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006850
CAS nummer: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,02562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4f2b4b1d8c967c55d5bdc79bf2abfbefe897711&vid=a4f2b4b1d8c967c55d5bdc79bf2abfbefe897711&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White powder to crystal
Purity (GC) min. 99.0 %
Melting point 68.0 to 73.0 deg-C
Solubility in Methanol almost transparency