Vitamin B12 (cyanocobalamin)
Leverancier: Chemicon
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CHMCCBL55EA
0
EUR
CHMCCBL55
OXBTCBL55
Vitamin B12 (cyanocobalamin)
Vitamin B12 (cyanocobalamin)
Formule:
C₆₃H₈₈CoN₁₄O₁₄P MW: 1355,38 g/mol Smeltpunt: 320 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00151092 CAS nummer: 68-19-9 EINECS: 200-680-0 Merck Index: 12,10152 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...