Vitamin B12 (cyanocobalamin), Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

6791-5EA 368 EUR
6791-5
Vitamin B12 (cyanocobalamin), Millipore®
Vitamin B12 (cyanocobalamin)
Formule: C₆₃H₈₈CoN₁₄O₁₄P
MW: 1355,38 g/mol
Smeltpunt: 320 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00151092
CAS nummer: 68-19-9
EINECS: 200-680-0
Merck Index: 12,10152

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR