Dicyclohexylamine 99+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: DCHA
181050025 181050050 181055000 181051000
ACRO181050025EA 115 EUR
ACRO181050025 ACRO181050050 ACRO181055000 ACRO181051000
Dicyclohexylamine 99+%
Dicyclohexylamine
Formule: C₁₂H₂₃N
Kookpunt: 256 °C (1013 hPa)
Smeltpunt: –2 °C
Dichtheid: 0,91 g/cm³
Vlampunt: 103 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011658
CAS nummer: 101-83-7
EINECS: 202-980-7
UN: 2565
ADR: 8,III
Merck Index: 13,03122
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4850 (20°C, 589 nm)
GC ≥99.0 %