2,6-Dihydroxy-9,10-anthraquinone
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2,6-Dihydroxyanthracene-9,10-dione, Anthraflavic acid, 2,6-Dihydroxyanthraquinone
SB01965.1 SB01965EA SB01965EE
MAYBSB01965.1EA 0 EUR
MAYBSB01965.1 MAYBSB01965EA MAYBSB01965EE
2,6-Dihydroxy-9,10-anthraquinone
2,6-Dihydroxy-9,10-anthraquinone
Formule: C₁₄H₈O₄
MW: 240.22 g/mol
Smeltpunt: > 330 °C
MDL Nummer: MFCD00001228
CAS nummer: 84-60-6
EINECS: 201-544-3