Dibenzofuran for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.20408.0005 8.20408.0250
8.20408.0005EA 32.1 EUR
8.20408.0005 8.20408.0250
Dibenzofuran for synthesis, Sigma-Aldrich®
Dibenzofuran
Formule: C₁₂H₈O
Kookpunt: 285 °C (1013 hPa)
Smeltpunt: 79…82 °C
Dichtheid: 0,966 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004968
CAS nummer: 132-64-9
EINECS: 205-071-3
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 79 - 82 °C
Molar Mass 168.19 g/mol
Boiling Point 287 - 289 °C
Flash Point 148 °C
Density 1.3 g/cm3 (20 °C)
Synonyms Diphenylene oxide
Hill Formula C12H8O