Fluorescein
Leverancier: Chemicon
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
MAB045
MAB045A
MAB045P
MAB045X
CHMCMAB045EA
0
EUR
CHMCMAB045
CHMCMAB045A
CHMCMAB045P
CHMCMAB045X
Fluorescein
Fluorescein
Formule:
C₂₀H₁₂O₅ Smeltpunt: 314…316 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00005050 CAS nummer: 2321-07-5 EINECS: 219-031-8 Merck Index: 13,04185 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...