Ethylene carbonate ≥99%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 99%
CAS No.: 96-49-1
Molecular Formula: C3H4O3
Formula Weight: 88.06
Flash Point: 160°(320°F)
Melting Point: 35-37°
Boiling Point: 244-245°
Solubility: Miscible (40%) with water, alchohol ethyl acetate, benzene and chloroform. Soluble in ether, n-Butanol, and carbon tetrachloride.
Beilstein Reference: 106249
MDL No.: MFCD00005382
Formule:
C₃H₄O₃ Kookpunt: 244…245 °C Smeltpunt: 35…37 °C Dichtheid: 1,320 Vlampunt: 160 °C (320 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00005382 CAS nummer: 96-49-1 EINECS: 202-510-0 |