5-Nitro-2-furaldehyde 97%
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-Nitrofurfural
AC17386DA
AC17386.10
AC17386.50
MAYBAC17386DAEA
0
EUR
MAYBAC17386DA
MAYBAC17386.10
MAYBAC17386.50
5-Nitro-2-furaldehyde 97%
5-Nitro-2-furaldehyde
Formule:
C₅H₃NO₄ Kookpunt: 272 °C (1013 hPa) Smeltpunt: 34…35 °C Dichtheid: 1,34 g/cm³ Vlampunt: 33 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003230 CAS nummer: 698-63-5 EINECS: 211-816-3 UN: 1325 ADR: 4.1,III |
Specification Test Results
Appearance | Yellow to brown Low melting crystalline solid |
Infrared spectrum | Conforms |
Melting point | 33 to 37 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...