5-Nitro-2-furaldehyde ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 5-Nitrofurfural
A10618.06 A10618.14 A10618.18
A10618.06EA 31 EUR
A10618.06 A10618.14 A10618.18
5-Nitro-2-furaldehyde ≥99%
5-Nitro-2-furaldehyde
Purity: 99% CAS No.: 698-63-5 Molecular Formula: C5H3NO4 Formula Weight: 141.09 Flash Point: 100°(212°F) Refractive Index: 1.5900 Melting Point: 34-38° Boiling Point: 228° Beilstein Reference: 120539 MDL No.: MFCD00003230
Formule: C₅H₃NO₄
Kookpunt: 228 °C
Smeltpunt: 34…38 °C
Dichtheid: 1,349
Vlampunt: 33 °C (91 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003230
CAS nummer: 698-63-5
EINECS: 211-816-3
UN: 1325
ADR: 4.1,III
Order Now
Specificaties
Environmentally Preferable