1,3-Difenylguanidine ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A15234.0B A15234.22 A15234.30
A15234.0BEA 99.3 EUR
A15234.0B A15234.22 A15234.30
1,3-Difenylguanidine ≥97%
1,3-Difenylguanidine

Purity: 97% CAS No.: 102-06-7 Molecular Formula: C13H13N3 Formula Weight: 211.27 Flash Point: 170°(338°F) Odour: Characteristic Boiling Point: 170° dec. Solubility: Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids Beilstein Reference: 1875653 Merck Reference: 14,3324 UN No.: 3077 MDL No.: MFCD00001758

Formule: C₁₃H₁₃N₃
Kookpunt: >170 °C (ontbinding)
Smeltpunt: 146…150 °C
Dichtheid: 1,130
Vlampunt: 170 °C (338 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001758
CAS nummer: 102-06-7
EINECS: 203-002-1
UN: 3077
ADR: 9,III
Merck Index: 14,3324

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR