Potassium tetrahydroborate 98+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potassium tetrahydridoborate, Potassium borohydride
196551000 196555000 196550025 196550050
ACRO196551000EA 90.4 EUR
ACRO196551000 ACRO196555000 ACRO196550025 ACRO196550050
Potassium tetrahydroborate 98+%
Potassium tetrahydroborate
Formule: H₄BK
MW: 53.94 g/mol
Smeltpunt: 500
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011396
CAS nummer: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3 I
Merck Index: 12,07777
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration ≥98.0 %