Cesium bromide, ultra dry ≥99.9% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Cesiumbromide
Ampuled under argon
14289.09
14289.18
14289.09EA
159
EUR
14289.09
14289.18
Cesium bromide, ultra dry ≥99.9% (metals basis)
Cesium bromide
Purity: 99.9%
CAS No.: 7787-69-1
Molecular Formula: CsBr
Formula Weight: 212.81
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.44
Refractive Index: 1.6984
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 636°
Boiling Point: 1300°
Merck Reference: 14,2009
MDL No.: MFCD00010954
Formule:
BrCs MW: 212.81 g/mol Kookpunt: 1300 °C Smeltpunt: 636 Dichtheid: 4.44 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010954 CAS nummer: 7787-69-1 EINECS: 232-130-0 Merck Index: 14,2009 |