Kaliumperchloraat, anhydrous 99,0-100,5 ACS
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
11630.22
11630.36
11630.22EA
44.8
EUR
11630.22
11630.36
Kaliumperchloraat, anhydrous 99,0-100,5 ACS
Kaliumperchloraat
Purity: 99.0-100.5%
CAS No.: 7778-74-7
Molecular Formula: KClO4
Formula Weight: 138.55
Physical Form: -100 Mesh Powder
Density: 2.52
Melting Point: 400° dec.
Solubility: Partially soluble in water. Insoluble in alcohol.
Merck Reference: 14,7653
UN No.: 1489
MDL No.: MFCD00011362
For Parr Bomb Work
Formule:
ClKO₄ MW: 138.55 g/mol Smeltpunt: 400 °C (ontbinding) Dichtheid: 2.52 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011362 CAS nummer: 7778-74-7 EINECS: 231-912-9 UN: 1489 ADR: 5.1 II Merck Index: 14,7653 |