Sodium hydrogen carbonate
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Sodium bicarbonate
8551.1
8551.2
8551.5
ROTH8551.1EA
0
EUR
ROTH8551.1
ROTH8551.2
ROTH8551.5
Sodium hydrogen carbonate
Sodium hydrogen carbonate
Formule:
NaHCO₃ Smeltpunt: 270 °C Dichtheid: 2,159 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003528 CAS nummer: 144-55-8 EINECS: 205-633-8 Merck Index: 13,08655 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...