Cesium iodide ≥99.999% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Cesiumiodide
10992.14
10992.22
10992.14EA
114
EUR
10992.14
10992.22
Cesium iodide ≥99.999% (metals basis)
Cesium iodide
Purity: 99.999%
CAS No.: 7789-17-5
Molecular Formula: CsI
Formula Weight: 259.81
Physical Form: Crystalline
Refractive Index: 1.7876
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 626°
Boiling Point: 1280°
Merck Reference: 14,2014
MDL No.: MFCD00010962
Formule:
CsI MW: 259.81 g/mol Kookpunt: 1280 °C Smeltpunt: 626 °C Dichtheid: 3.21 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010962 CAS nummer: 7789-17-5 EINECS: 232-145-2 Merck Index: 14,2014 |