Potassium iodate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P751.1 P751.2
ROTHP751.1EA 57.9 EUR
ROTHP751.1 ROTHP751.2
Potassium iodate
Potassium iodate
Formule: KlO₃
Smeltpunt: 560 °C
Dichtheid: 3,98 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011406
CAS nummer: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 13,07726

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR