Potassium periodate 99.5%, extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potassium tetroxoiodate (VII), Potassium metaperiodate
198391000 198395000 198390025
ACRO198391000EA 88.8 EUR
ACRO198391000 ACRO198395000 ACRO198390025
Potassium periodate 99.5%, extra pure
Potassium periodate
Formule: KlO₄
Smeltpunt: 582 °C
Dichtheid: 3,62 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011407
CAS nummer: 7790-21-8
EINECS: 232-196-0
UN: 1479
ADR: 5.1,II
Merck Index: 13,07739
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Titration Iodometric ≥99.4 %
Other halogens (as Cl) ≤0.01 %
Manganese (Mn) ≤1 ppm