Tripotassium orthophosphate, anhydrous 97%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potassium phosphate tribasic, Potassium phosphate
387680250 387685000 387680010 387680025 387680100
ACRO387680250EA 30.2 EUR
ACRO387680250 ACRO387685000 ACRO387680010 ACRO387680025 ACRO387680100
Tripotassium orthophosphate, anhydrous 97%, pure
Tripotassium orthophosphate
Formule: K₃O₄P
MW: 212.27 g/mol
Smeltpunt: 1340 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036295
CAS nummer: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830
Order Now
Specification Test Results
Appearance White Powder or granules and/or chunks
Titration excess HCl ≥96.0 %