, anhydrous, EMPROVE® EXPERT BP, Ph. Eur., USP for biopharmaceutical production, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
1.37010.5000 1.37010.9012 1.37010.9029
1.37010.5000EA 675 EUR
1.37010.5000 1.37010.9012 1.37010.9029
, anhydrous, EMPROVE® EXPERT BP, Ph. Eur., USP for biopharmaceutical production, SAFC®
di-Potassium hydrogen phosphate
Formule: K₂HPO₄
MW: 174.18 g/mol
Smeltpunt: 340
Dichtheid: 2.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011383
CAS nummer: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM