Disodium hydrogen phosphate, anhydrous 98+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
A11817.0C A11817.0E A11817.30 A11817.36 A11817.A1 A11817.A7
A11817.0CEA 344 EUR
A11817.0C A11817.0E A11817.30 A11817.36 A11817.A1 A11817.A7
Disodium hydrogen phosphate, anhydrous 98+%
Disodium hydrogen phosphate
Purity: 98+% CAS No.: 7558-79-4 Molecular Formula: HNa2O4P Formula Weight: 141.98 Odour: Odourless Merck Reference: 14,8659 MDL No.: MFCD00003496
Formule: Na₂HPO₄
MW: 141.96 g/mol
Smeltpunt: 250
Dichtheid: 1.53 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003496
CAS nummer: 7558-79-4
EINECS: 231-448-7
Bereik: 01-2119489797-11
Merck Index: 14,8659