Potassium dihydrogen phosphate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
3904.1 3904.2 3904.3 3904.5
ROTH3904.1EA 0 EUR
ROTH3904.1 ROTH3904.2 ROTH3904.3 ROTH3904.5
Potassium dihydrogen phosphate
Potassium dihydrogen phosphate
Formule: KH₂PO₄
Kookpunt: ∼100 °C (1013 hPa)
Smeltpunt: 253 °C
Dichtheid: 2,3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011401
CAS nummer: 7778-77-0
EINECS: 231-913-4
Bereik: 01-2119490224-41
Merck Index: 13,07744