Potassium disulphite, EMPROVE® ESSENTIAL E224, Ph. Eur., NF, extra pure suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Dipotassium disulphite, Potassium pyrosulphite, Potassium metabisulphite, di-Potassium disulphite
1.05056.5000 1.05056.9029
1.05056.5000EA 1140 EUR
1.05056.5000 1.05056.9029
Potassium disulphite, EMPROVE® ESSENTIAL E224, Ph. Eur., NF, extra pure suitable for use as excipient, SAFC®
Potassium disulphite
Formule: K₂O₅S₂
MW: 222.33 g/mol
Smeltpunt: 190
Dichtheid: 1.2 g/cm³ (20 °C)
MDL Nummer: MFCD00167605
CAS nummer: 16731-55-8
EINECS: 240-795-3
Merck Index: 13,07729