Cesium sulphate 99+%, pure
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
218700250
218701000
218700100
218702500
ACRO218700250EA
35.9
EUR
ACRO218700250
ACRO218701000
ACRO218700100
ACRO218702500
Cesium sulphate 99+%, pure
Cesium sulphate
Formule:
Cs₂O₄S Kookpunt: 1900 °C (760 mmHg) Smeltpunt: 1005 °C Dichtheid: 4,24 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010959 CAS nummer: 10294-54-9 EINECS: 233-662-6 Merck Index: 13,02025 |
Specification Test Results
Appearance | White Crystalline powder |
Assay | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...