Potassium sulphate 99.0-101.0% (alkalimetric, calculated on anhydrous substance), EMPROVE® ESSENTIAL BP, Ph. Eur., JP, extra pure suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.05150.1000 1.05150.9029
1.05150.1000EA 1390 EUR
1.05150.1000 1.05150.9029
Potassium sulphate 99.0-101.0% (alkalimetric, calculated on anhydrous substance), EMPROVE® ESSENTIAL BP, Ph. Eur., JP, extra pure suitable for use as excipient, SAFC®
Potassium sulphate
Formule: K₂SO₄
Kookpunt: 1698 °C (1013 hPa)
Smeltpunt: 1069 °C
Dichtheid: 2,66 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011388
CAS nummer: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759