Potassium sulphate
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P022.1
P022.2
ROTHP022.1EA
59.9
EUR
ROTHP022.1
ROTHP022.2
Potassium sulphate
Potassium sulphate
Formule:
K₂SO₄ MW: 174.26 g/mol Kookpunt: 1698 Smeltpunt: 1069 Dichtheid: 2.66 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011388 CAS nummer: 7778-80-5 EINECS: 231-915-5 Merck Index: 13,07759 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...