Lithium wolframate ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Lithium tungsten oxide, Lithium tungstate
43366.30
43366.18
43366.30EA
429
EUR
43366.30
43366.18
Lithium wolframate ≥99%
Lithium wolframate
Purity: 99% min
CAS No.: 13568-45-1
Molecular Formula: Li2WO4
Formula Weight: 261.73
Physical Form: Powder
Density: 3.71
Melting Point: 742°
MDL No.: MFCD00016185
Formule:
Li₂O₄W MW: 261.72 g/mol Dichtheid: 3.71 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00016185 CAS nummer: 13568-45-1 EINECS: 236-978-2 |