Calciumchloride, anhydrous ≥95%, white powder
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Chloro calcium, Calcium dichloride, CaCl2
0215350291 0215350280 0215350290
ICNA0215350291EA 140 EUR
ICNA0215350291 ICNA0215350280 ICNA0215350290
Calciumchloride, anhydrous ≥95%, white powder
Calciumchloride
Anhydrous Calcium Chloride is highly hygroscopic and is often used as a desiccant. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability.
Soluble in water with heat liberation, ethanol, acetic acid, acetone.
Formule: CaCl₂
MW: 110.98 g/mol
Kookpunt: 1600
Smeltpunt: 772
Dichtheid: 2.174 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010903
CAS nummer: 10043-52-4
EINECS: 233-140-8
Bereik: 01-2119494219-28
Merck Index: 14,01659