Bariumperchloraat ≥97.0%, AnalaR NORMAPUR® analytical reagent
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
21749.236EA
206
EUR
21749.236
Bariumperchloraat ≥97.0%, AnalaR NORMAPUR® analytical reagent
Bariumperchloraat
Formule:
Ba(ClO₄)₂ MW: 336,23 g/mol Smeltpunt: 505 °C Dichtheid: 3,2 g/cm³ (20 °C) Vlampunt: 21 °C (gesloten beker) |
MDL Nummer:
MFCD00003440 CAS nummer: 13465-95-7 EINECS: 236-710-4 UN: 1447 ADR: 5.1 (6.1),II Merck Index: 12,01016 |
Specification Test Results
Assay | Min. 97.0 % |
Heavy metals (as Pb) | Max. 5 ppm |
Insolubility in methanol | Max. 0.1 % |
Not precipitated by H2SO4 (as SO4) | Max. 0.2 % |
Water | Max. 4 % |
Cl (Chloride) | Max. 100 ppm |
Ca (Calcium) | Max. 0.05 % |
Fe (Iron) | Max. 5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...