Magnesium perchlorate ACS desiccant
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
413465000 413461000
ACRO413465000EA 319 EUR
ACRO413465000 ACRO413461000
Magnesium perchlorate ACS desiccant
Magnesium perchlorate
Formule: Cl₂MgO₈
Smeltpunt: 250 °C
Dichtheid: 2,21 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011102
CAS nummer: 10034-81-8
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II
Merck Index: 13,05702
Order Now
Specification Test Results
Appearance White Crystalline powder or flakes and/or chunks
Loss on drying ≤8 % (1 g, 190°C)
Suitability (for moisture absorption) Passes test
Titrable base ≤0.025 meq/g
Titrable free acid ≤0.005 meq/g