Calcium carbonate 99+% ACS
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Marble
423515000
423510025
423511000
ACRO423515000EA
65.5
EUR
ACRO423515000
ACRO423510025
ACRO423511000
Calcium carbonate 99+% ACS
Calcium carbonate
Formule:
CaCO₃ Dichtheid: 2,71 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010906 CAS nummer: 471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Specification Test Results
Appearance | White Crystalline powder |
Titration Complexometric | ≥99.0 % (on dried substance) |
Chloride (Cl) | ≤0.001 % |
Fluoride (F) | ≤0.0015 % |
Sulfate (SO4) | ≤0.01 % |
Ammonium (NH4) | ≤0.003 % |
Barium (Ba) | ≤0.01 % |
Iron (Fe) | ≤0.003 % |
Magnesium (Mg) | ≤0.02 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.1 % |
Strontium (Sr) | ≤0.1 % |
Heavy metals (ICP-OES) | ≤0.001 % |
Insoluble matter | ≤0.01 % (in dilute HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...