Magnesium hydroxide carbonate light BP, Ph. Eur., extra pure, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic
1.05828.9015 1.05828.9020
1.05828.9015EA 0 EUR
1.05828.9015 1.05828.9020
Magnesium hydroxide carbonate light BP, Ph. Eur., extra pure, SAFC®
Magnesium hydroxide carbonate
Formule: MgCO₃ (linear)
MW: 84.31 g/mol
Kookpunt: 3600
Dichtheid: 2.16 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149783
CAS nummer: 12125-28-9
EINECS: 235-192-7