Calcium fluoride, ultra dry ≥99.99% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Ampuled under argon
35742.06
35742.14
35742.22
35742.06EA
128
EUR
35742.06
35742.14
35742.22
Calcium fluoride, ultra dry ≥99.99% (metals basis)
Calcium fluoride
Purity: 99.99%
CAS No.: 7789-75-5
Molecular Formula: CaF2
Formula Weight: 78.08
Physical Form: Powder, Ampouled under argon
Density: 3.18
Refractive Index: 1.434
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 1403°
Boiling Point: ca 2500°
Merck Reference: 14,1667
MDL No.: MFCD00010907
Formule:
CaF₂ Kookpunt: ∼2500 °C Smeltpunt: 1403 °C Dichtheid: 3,18 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010907 CAS nummer: 7789-75-5 EINECS: 232-188-7 Merck Index: 14,1667 |