Calcium fluoride 99
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A10913.0E A10913.22 A10913.36
A10913.0EEA 327 EUR
A10913.0E A10913.22 A10913.36
Calcium fluoride 99
Calcium fluoride
Purity: 99% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Refractive Index: 1.434 Odour: Odourless Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907
Formule: CaF₂
MW: 78.07 g/mol
Kookpunt: ∼2500 °C
Smeltpunt: 1403 °C
Dichtheid: 3.18
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010907
CAS nummer: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669