Magnesium oxide substrate 10x10x0.5mm, polished one side 111 orientation
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Magnesia usta
40273.KT 40273.KU
40273.KTEA 127 EUR
40273.KT 40273.KU
Magnesium oxide substrate 10x10x0.5mm, polished one side 111 orientation
Magnesium oxide
Molecular Formula: MgO MDL No.: MFCD00011109
Formule: MgO
MW: 40.3 g/mol
Kookpunt: 3600
Smeltpunt: 2800
Dichtheid: 3.58 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011109
CAS nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700