Magnesium oxide, 2 part ceramic adhesive paste
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43135.A4 43135.36
43135.A4EA 508 EUR
43135.A4 43135.36
Magnesium oxide, 2 part ceramic adhesive paste
Kleefstoffen
High temperature adhesive for bonding ceramic to metal.
High temperature adhesive for bonding ceramic to metal.
Mix ratio (w/w): 1.5:1 powder to liquid.
Cure conditions: 1-4 hours at room temperature, 2 hours at 94 °C
High temperature adhesive for bonding ceramic to metal